2-Chloro-N-fmoc-L-phenylalanine

Supplier: Matrix Scientific

Synonyms: Fmoc-Phe(2-Cl)-OH

041924-1G 041924-5G
101853-750EA 38.46 USD
101853-750 101853-752
2-Chloro-N-fmoc-L-phenylalanine
2-Chloro-N-fmoc-L-phenylalanine

Hazard: Irritant

Formula: C₂₄H₂₀ClNO₄
MW: 421.877 g/mol
MDL Number: MFCD00797578
CAS Number: 198560-41-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR