2-Bromo-N-fmoc-L-phenylalanine

Supplier: Matrix Scientific

Synonyms: Fmoc-Phe(2-Br)-OH

041922-1G 041922-5G
101853-742EA 412.31 USD
101853-742 101853-744
2-Bromo-N-fmoc-L-phenylalanine
2-Bromo-N-fmoc-L-phenylalanine

Hazard: Irritant

Formula: C₂₄H₂₀BrNO₄
MW: 466.331 g/mol
MDL Number: MFCD01311743
CAS Number: 220497-47-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR