Fmoc-2-bromo-D-phenylalanine
Supplier: Matrix Scientific
Synonyms:
(R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(2-bromophenyl)propanoic acid, Fmoc-D-Phe(2-Br)-OH
041921-1G
041921-5G
101853-738EA
412.31
USD
101853-738
101853-740
Fmoc-2-bromo-D-phenylalanine
Fmoc-2-bromo-D-phenylalanine
Hazard: Irritant
Formula:
C₂₄H₂₀BrNO₄ MW: 466.34 g/mol |
MDL Number:
MFCD01311770 CAS Number: 220497-79-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...