Fmoc-2,4-dichloro-D-phenylalanine

Supplier: Matrix Scientific

Synonyms: 2,4-Dichloro-D-phenylalanine, N-FMOC protected

041919-1G 041919-5G
101853-730EA 547.69 USD
101853-730 101853-732
Fmoc-2,4-dichloro-D-phenylalanine
Fmoc-2,4-dichloro-D-phenylalanine

Hazard: Irritant

Formula: C₂₄H₁₉Cl₂NO₄
MW: 456.33 g/mol
MDL Number: MFCD01863047
CAS Number: 352351-61-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR