Fmoc-2-(trifluoromethyl)-D-phenylalanine
Supplier: Matrix Scientific
Synonyms:
(R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(2-(trifluoromethyl)phenyl)propanoic acid, 2-(Trifluoromethyl)-D-phenylalanine, N-FMOC protected, Fmoc-D-Phe(2-CF₃)-OH
041917-1G
041917-5G
101853-722EA
1095.38
USD
101853-722
101853-724
Fmoc-2-(trifluoromethyl)-D-phenylalanine
Fmoc-2-(trifluoromethyl)-D-phenylalanine
Hazard: Irritant
Formula:
C₂₅H₂₀F₃NO₄ MW: 455.44 g/mol |
MDL Number:
MFCD01863051 CAS Number: 352523-15-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...