Fmoc-D-Tic-OH
Supplier: Matrix Scientific
Synonyms:
Fmoc-D-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid
041915-1G
041915-5G
101853-714EA
101.54
USD
101853-714
101853-716
Fmoc-D-Tic-OH
Fmoc-D-Tic-OH
Hazard: Irritant
Formula:
C₂₅H₂₁NO₄ MW: 399.446 g/mol |
MDL Number:
MFCD00144345 CAS Number: 130309-33-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...