(S)-2-((tert-Butoxycarbonyl)amino)-3-(4-methoxyphenyl)propanoic acid ≥95%
Supplier: Matrix Scientific
Synonyms:
Boc-Tyr(Me)-OH, N-Boc-O-methyl-L-tyrosine
041914-1G
041914-5G
101853-710EA
84.62
USD
101853-710
101853-712
(S)-2-((tert-Butoxycarbonyl)amino)-3-(4-methoxyphenyl)propanoic acid ≥95%
(S)-2-((tert-Butoxycarbonyl)amino)-3-(4-methoxyphenyl)propanoic acid
Hazard: Irritant
Formula:
C₁₅H₂₁NO₅ MW: 295.3 g/mol |
MDL Number:
MFCD00065603 CAS Number: 53267-93-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...