Boc-D-Tyr(ME)-OH ≥95%

Supplier: Matrix Scientific

Synonyms: Boc-D-Phe(4-OMe)-OH, N-Boc-4-methoxy-D-phenylalanine, N-(tert-Butoxycarbonyl)-4-methoxy-D-phenylalanine

041913-1G 041913-5G
101853-706EA 61.54 USD
101853-706 101853-708
Boc-D-Tyr(ME)-OH ≥95%
Boc-D-Tyr(ME)-OH

Hazard: Irritant

Formula: C₁₅H₂₁NO₅
MW: 295.335 g/mol
MDL Number: MFCD00065604
CAS Number: 68856-96-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR