(S)-2-((tert-Butoxycarbonyl)amino)-3-(4-ethoxyphenyl)propanoic acid

Supplier: Matrix Scientific

Synonyms: Boc-Tyr(Et)-OH

041912-1G 041912-5G
101853-702EA 21.54 USD
101853-702 101853-704
(S)-2-((tert-Butoxycarbonyl)amino)-3-(4-ethoxyphenyl)propanoic acid
(S)-2-((tert-Butoxycarbonyl)amino)-3-(4-ethoxyphenyl)propanoic acid

Hazard: Irritant

Formula: C₁₆H₂₃NO₅
MW: 309.3575 g/mol
MDL Number: MFCD00065601
CAS Number: 76757-91-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR