Boc-L-thyroxine

Supplier: Matrix Scientific

041910-1G 041910-5G
101853-694EA 686.15 USD
101853-694 101853-696
Boc-L-thyroxine
Boc-L-thyroxine

Hazard: Irritant

Formula: C₂₀H₁₉I₄NO₆
MDL Number: MFCD02682290
CAS Number: 88404-22-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR