Boc-L-thyronine

Supplier: Matrix Scientific

041909-1G 041909-5G
101853-690EA 1095.38 USD
101853-690 101853-692
Boc-L-thyronine
Boc-L-thyronine

Hazard: Irritant

Formula: C₂₀H₂₃NO₆
MDL Number: MFCD02682294
CAS Number: 1213135-25-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR