(S)-2-((tert-Butoxycarbonyl)amino)-4-phenylbutanoic acid

Supplier: Matrix Scientific

041908-5G 041908-25G
101853-686EA 29.23 USD
101853-686 101853-688
(S)-2-((tert-Butoxycarbonyl)amino)-4-phenylbutanoic acid
(S)-2-((tert-Butoxycarbonyl)amino)-4-phenylbutanoic acid

Hazard: Irritant

Formula: C₁₅H₂₁NO₄
MDL Number: MFCD00076904
CAS Number: 100564-78-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR