Boc-L-3,3-diphenylalanine
Supplier: Matrix Scientific
Synonyms:
(S)-2-((tert-Butoxycarbonyl)amino)-3,3-diphenylpropanoic acid
041907-1G
041907-5G
101853-680EA
326.15
USD
101853-680
101853-682
Boc-L-3,3-diphenylalanine
Boc-L-3,3-diphenylalanine
Hazard: Irritant
Formula:
C₂₀H₂₃NO₄ MW: 341.41 g/mol |
MDL Number:
MFCD00191186 CAS Number: 138662-63-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...