Boc-D-thyroxine

Supplier: Matrix Scientific

041906-1G 041906-5G
101853-676EA 823.08 USD
101853-676 101853-678
Boc-D-thyroxine
Boc-D-thyroxine

Hazard: Irritant

Formula: C₂₀H₁₉I₄NO₆
MDL Number: MFCD02682291
CAS Number: 89624-64-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR