Boc-DL-thyronine

Supplier: Matrix Scientific

041905-1G 041905-5G
101853-672EA 1095.38 USD
101853-672 101853-674
Boc-DL-thyronine
Boc-DL-thyronine

Hazard: Irritant

Formula: C₂₀H₂₃NO₆
MDL Number: MFCD02682676
CAS Number: 1452572-11-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR