Boc-DL-o-tyrosine
Supplier: Matrix Scientific
041904-1G
041904-5G
101853-668EA
1095.38
USD
101853-668
101853-670
Boc-DL-o-tyrosine
Boc-DL-o-tyrosine
Hazard: Irritant
Formula:
C₁₄H₁₉NO₅ MW: 281.31 g/mol |
MDL Number:
MFCD02682299 CAS Number: 203569-04-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...