Boc-DL-m-tyrosine

Supplier: Matrix Scientific

041903-1G 041903-5G
101853-664EA 684.62 USD
101853-664 101853-666
Boc-DL-m-tyrosine
Boc-DL-m-tyrosine

Hazard: Irritant

Formula: C₁₄H₁₉NO₅
MDL Number: MFCD02682297
CAS Number: 174732-96-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR