Boc-D-homophenylalanine

Supplier: Matrix Scientific

Synonyms: Boc-D-Homophe-OH, (R)-2-(Boc-amino)-4-phenylbutyric acid, Boc-D-HoPhe-OH

041902-1G 041902-5G
101853-660EA 21.54 USD
101853-660 101853-662
Boc-D-homophenylalanine
Boc-D-homophenylalanine

Hazard: Irritant

Formula: C₁₅H₂₁NO₄
MW: 279.336 g/mol
MDL Number: MFCD00076905
CAS Number: 82732-07-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR