N-Boc-β-phenyl-D-phenylalanine

Supplier: Matrix Scientific

Synonyms: N-(tert-Butoxycarbonyl)-beta-phenyl-D-phenylalanine, Boc-D-Ala(3,3-diphenyl)-OH, Boc-D-3,3-diphenylalanine

041901-1G 041901-5G
101853-656EA 326.15 USD
101853-656 101853-658
N-Boc-β-phenyl-D-phenylalanine
N-Boc-beta-phenyl-D-phenylalanine

Hazard: Irritant

Formula: C₂₀H₂₃NO₄
MW: 341.40 g/mol
MDL Number: MFCD00191187
CAS Number: 143060-31-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR