Boc-β-methyl-DL-phenylalanine

Supplier: Matrix Scientific

041900-100MG 041900-1G
101853-652EA 138.46 USD
101853-652 101853-654
Boc-β-methyl-DL-phenylalanine
Boc-beta-methyl-DL-phenylalanine

Hazard: Irritant

Formula: C₁₅H₂₁NO₄
MW: 279.34 g/mol
MDL Number: MFCD01823263
CAS Number: 630424-76-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR