Boc-α-methyl-L-phenylalanine

Supplier: Matrix Scientific

Synonyms: (R)-2-((tert-Butoxycarbonyl)amino)-2-methyl-3-phenylpropanoic acid, Boc-α-Me-Phe-OH

041899-250MG 041899-1G
101853-648EA 89.23 USD
101853-648 101853-650
Boc-α-methyl-L-phenylalanine
Boc-alpha-methyl-L-phenylalanine

Hazard: Irritant

Formula: C₁₅H₂₁NO₄
MW: 279.34 g/mol
MDL Number: MFCD02682284
CAS Number: 111771-58-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR