Boc-α-methyl-D-phenylalanine
Supplier: Matrix Scientific
041898-250MG
041898-1G
041898-5G
101853-642EA
723.08
USD
101853-642
101853-644
101853-646
Boc-α-methyl-D-phenylalanine
Boc-alpha-methyl-D-phenylalanine
Hazard: Irritant
Formula:
C₁₅H₂₁NO₄ MW: 279.34 g/mol |
MDL Number:
MFCD02682673 CAS Number: 53940-88-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...