Boc-α-methyl-3-methoxy-DL-phenylalanine

Supplier: Matrix Scientific

041897-100MG 041897-500MG
101853-638EA 275.38 USD
101853-638 101853-640
Boc-α-methyl-3-methoxy-DL-phenylalanine
Boc-alpha-methyl-3-methoxy-DL-phenylalanine

Hazard: Irritant

Formula: C₁₆H₂₃NO₅
MW: 309.37 g/mol
MDL Number: MFCD02682276
CAS Number: 1185301-00-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR