Boc-7-hydroxy-D-Tic-OH

Supplier: Matrix Scientific

Synonyms: Boc-D-7-hydroxy-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid, (R)-2-(tert-Butoxycarbonyl)-7-hydroxy-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid

041895-250MG 041895-1G
101853-630EA 152.31 USD
101853-630 101853-632
Boc-7-hydroxy-D-Tic-OH
Boc-7-hydroxy-D-Tic-OH

Hazard: Irritant

Formula: C₁₅H₁₉NO₅
MW: 293.319 g/mol
MDL Number: MFCD00792389
CAS Number: 214630-00-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR