Boc-5-bromo-2-methoxy-L-phenylalanine

Supplier: Matrix Scientific

041894-500MG 041894-1G
101853-626EA 598.46 USD
101853-626 101853-628
Boc-5-bromo-2-methoxy-L-phenylalanine
Boc-5-bromo-2-methoxy-L-phenylalanine

Hazard: Irritant

Formula: C₁₅H₂₀BrNO₅
MW: 374.24 g/mol
MDL Number: MFCD01320847
CAS Number: 261165-03-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR