Boc-5-bromo-2-methoxy-D-phenylalanine

Supplier: Matrix Scientific

041893-500MG 041893-1G
101853-622EA 686.15 USD
101853-622 101853-624
Boc-5-bromo-2-methoxy-D-phenylalanine
Boc-5-bromo-2-methoxy-D-phenylalanine

Hazard: Irritant

Formula: C₁₅H₂₀BrNO₅
MDL Number: MFCD01320878
CAS Number: 261380-17-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR