N-(tert-Butoxycarbonyl)-4-nitro-D-phenylalanine

Supplier: Matrix Scientific

Synonyms: N-Boc-4-nitro-D-phenylalanine, Boc-D-Phe(4-NO2)-OH, Boc-D-Phe(4-NO₂)-OH

041891-1G 041891-5G
101853-614EA 15.38 USD
101853-614 101853-616
N-(tert-Butoxycarbonyl)-4-nitro-D-phenylalanine
N-(tert-Butoxycarbonyl)-4-nitro-D-phenylalanine

Hazard: Irritant

Formula: C₁₄H₁₈N₂O₆
MW: 310.31 g/mol
Melting Pt: 120 °C
MDL Number: MFCD00069880
CAS Number: 61280-75-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR