N-Boc-4-methyl-L-phenylalanine
Supplier: Matrix Scientific
041890-1G
041890-5G
101853-610EA
187.69
USD
101853-610
101853-612
N-Boc-4-methyl-L-phenylalanine
Hazard: Irritant
Formula:
C₁₅H₂₁NO₄ |
MDL Number:
MFCD01317027 CAS Number: 80102-26-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...