Boc-Phe(4-CN)-OH

Supplier: Matrix Scientific

Synonyms: Boc-4-cyano-Phe-OH, N-Boc-4-cyano-L-phenylalanine, Boc-Phe(4-CN)-OH, N-(tert-Butoxycarbonyl)-4-cyano-L-phenylalanine

041886-1G 041886-5G
101853-594EA 158.46 USD
101853-594 101853-596
Boc-Phe(4-CN)-OH
Boc-Phe(4-CN)-OH

Hazard: Irritant

Formula: C₁₅H₁₈N₂O₄
MW: 290.319 g/mol
MDL Number: MFCD00672526
CAS Number: 131724-45-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR