N-(tert-Butoxycarbonyl)-4-chloro-L-phenylalanine ≥95%

Supplier: Matrix Scientific

Synonyms: Boc-Phe(4-Cl)-OH, N-Boc-4-chloro-L-phenylalanine

041884-1G 041884-5G
101853-586EA 29.23 USD
101853-586 101853-588
N-(tert-Butoxycarbonyl)-4-chloro-L-phenylalanine ≥95%
N-(tert-Butoxycarbonyl)-4-chloro-L-phenylalanine

Hazard: Irritant

Formula: C₁₄H₁₈ClNO₄
MW: 299.75 g/mol
MDL Number: MFCD00057808
CAS Number: 68090-88-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR