N-(tert-Butoxycarbonyl)-4-chloro-D-phenylalanine
Supplier: Matrix Scientific
Synonyms:
N-Boc-4-chloro-D-phenylalanine, Boc-D-Phe(4-Cl)-OH, (R)-N-BOC-4-Chlorophenylalanine
041883-1G
041883-5G
101853-582EA
138.46
USD
101853-582
101853-584
N-(tert-Butoxycarbonyl)-4-chloro-D-phenylalanine
N-(tert-Butoxycarbonyl)-4-chloro-D-phenylalanine
Hazard: Irritant
Formula:
C₁₄H₁₈ClNO₄ MW: 299.75 g/mol |
MDL Number:
MFCD00076978 CAS Number: 57292-44-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...