N-(tert-Butoxycarbonyl)-4-bromo-D-phenylalanine ≥95%
Supplier: Matrix Scientific
Synonyms:
Boc-D-Phe(4-Br)-OH, N-Boc-4-bromo-D-phenylalanine
041881-1G
041881-5G
101853-574EA
21.54
USD
101853-574
101853-576
N-(tert-Butoxycarbonyl)-4-bromo-D-phenylalanine ≥95%
N-(tert-Butoxycarbonyl)-4-bromo-D-phenylalanine
Hazard: Irritant
Formula:
C₁₄H₁₈BrNO₄ MW: 344.21 g/mol |
MDL Number:
MFCD00270359 CAS Number: 79561-82-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...