Boc-4-bis(2-chloroethyl)amino-L-phenylalanine

Supplier: Matrix Scientific

041880-100MG 041880-500MG
101853-570EA 326.15 USD
101853-570 101853-572
Boc-4-bis(2-chloroethyl)amino-L-phenylalanine
Boc-4-bis(2-chloroethyl)amino-L-phenylalanine

Hazard: Irritant

Formula: C₁₈H₂₆Cl₂N₂O₄
MW: 405.32 g/mol
MDL Number: MFCD02682258
CAS Number: 79145-92-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR