Boc-D-Bpa-OH
Supplier: Matrix Scientific
Synonyms:
Boc-p-Bz-D-Phe-OH
041878-250MG
041878-1G
101853-562EA
169.23
USD
101853-562
101853-564
Boc-D-Bpa-OH
Boc-D-Bpa-OH
Hazard: Irritant
Formula:
C₂₁H₂₃NO₅ MW: 369.417 g/mol |
MDL Number:
MFCD00237572 CAS Number: 117666-94-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...