Boc-4-(trifluoromethyl)-D-phenylalanine

Supplier: Matrix Scientific

Synonyms: 4-(Trifluoromethyl)-D-phenylalanine, N-BOC protected, (R)-2-((tert-Butoxycarbonyl)amino)-3-(4-(trifluoromethyl)phenyl)propanoic acid

041875-1G 041875-5G
101853-550EA 238.46 USD
101853-550 101853-552
Boc-4-(trifluoromethyl)-D-phenylalanine
Boc-4-(trifluoromethyl)-D-phenylalanine

Hazard: Irritant

Formula: C₁₅H₁₈F₃NO₄
MW: 333.31 g/mol
MDL Number: MFCD00797557
CAS Number: 82317-83-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR