Boc-4-(Fmoc-aminomethyl)-L-phenylalanine

Supplier: Matrix Scientific

041874-1G 041874-5G
101853-546EA 598.46 USD
101853-546 101853-548
Boc-4-(Fmoc-aminomethyl)-L-phenylalanine
Boc-4-(Fmoc-aminomethyl)-L-phenylalanine

Hazard: Irritant

Formula: C₃₀H₃₂N₂O₆
MDL Number: MFCD01317029
CAS Number: 170157-61-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR