Boc-4-(Fmoc-aminomethyl)-D-phenylalanine
Supplier: Matrix Scientific
041873-1G
041873-5G
101853-542EA
598.46
USD
101853-542
101853-544
Boc-4-(Fmoc-aminomethyl)-D-phenylalanine
Boc-4-(Fmoc-aminomethyl)-D-phenylalanine
Hazard: Irritant
Formula:
C₃₀H₃₂N₂O₆ MW: 516.6 g/mol |
MDL Number:
MFCD02094577 CAS Number: 215302-77-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...