Boc-3-nitro-L-phenylalanine

Supplier: Matrix Scientific

Synonyms: Boc-L-3-Nitrophenylalanine, Boc-Phe(3-NO₂)-OH, 3-Nitro-L-phenylalanine, N-BOC protected

041871-1G 041871-5G
101853-534EA 275.38 USD
101853-534 101853-536
Boc-3-nitro-L-phenylalanine
Boc-3-nitro-L-phenylalanine

Hazard: Irritant

Formula: C₁₄H₁₈N₂O₆
MW: 310.31 g/mol
MDL Number: MFCD01317033
CAS Number: 131980-29-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR