Boc-3-nitro-D-phenylalanine

Supplier: Matrix Scientific

041870-1G 041870-5G
101853-530EA 207.69 USD
101853-530 101853-532
Boc-3-nitro-D-phenylalanine
Boc-3-nitro-D-phenylalanine

Hazard: Irritant

Formula: C₁₄H₁₈N₂O₆
MDL Number: MFCD01317702
CAS Number: 158741-21-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR