N-Boc-3-methyl-D-phenylalanine

Supplier: Matrix Scientific

Synonyms: (R)-2-((tert-Butoxycarbonyl)amino)-3-(m-tolyl)propanoic acid

041868-1G 041868-5G
101853-522EA 187.69 USD
101853-522 101853-524
N-Boc-3-methyl-D-phenylalanine
N-Boc-3-methyl-D-phenylalanine

Hazard: Irritant

Formula: C₁₅H₂₁NO₄
MW: 279.336 g/mol
MDL Number: MFCD01862943
CAS Number: 114873-14-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR