Boc-3-methoxy-L-tyrosine(dicyclohexylammonium) salt ≥95%

Supplier: Matrix Scientific

041867-1G 041867-5G
101853-518EA 386.15 USD
101853-518 101853-520
Boc-3-methoxy-L-tyrosine(dicyclohexylammonium) salt ≥95%
Boc-3-methoxy-L-tyrosine(dicyclohexylammonium) salt

Hazard: Irritant

Formula: C₂₇H₄₄N₂O₆
MW: 492.66 g/mol
MDL Number: MFCD28099699
CAS Number: 53267-94-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR