Boc-3-iodo-L-tyrosine
Supplier: Matrix Scientific
041865-1G
041865-5G
101853-514EA
275.38
USD
101853-514
101853-516
Boc-3-iodo-L-tyrosine
Boc-3-iodo-L-tyrosine
Hazard: Irritant
Formula:
C₁₄H₁₈INO₅ MW: 407.21 g/mol |
MDL Number:
MFCD01632008 CAS Number: 71400-63-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...