Boc-3-iodo-L-phenylalanine ≥95%

Supplier: Matrix Scientific

Synonyms: Boc-Phe(3-I)-OH

041864-1G 041864-5G
101853-510EA 384.62 USD
101853-510 101853-512
Boc-3-iodo-L-phenylalanine ≥95%
Boc-3-iodo-L-phenylalanine

Hazard: Irritant

Formula: C₁₄H₁₈INO₄
MW: 391.21 g/mol
MDL Number: MFCD01860644
CAS Number: 273221-75-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR