(R)-N-Boc-3-fluorophenylalanine

Supplier: Matrix Scientific

Synonyms: Boc-D-Phe(3-F)-OH

041861-1G 041861-5G
101853-498EA 149.23 USD
101853-498 101853-500
(R)-N-Boc-3-fluorophenylalanine
(R)-N-Boc-3-fluorophenylalanine

Hazard: Irritant

Formula: C₁₄H₁₈FNO₄
MW: 283.3 g/mol
MDL Number: MFCD00672523
CAS Number: 114873-11-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR