Boc-3-fluoro-DL-tyrosine

Supplier: Matrix Scientific

041860-100MG 041860-1G
101853-494EA 164.62 USD
101853-494 101853-496
Boc-3-fluoro-DL-tyrosine
Boc-3-fluoro-DL-tyrosine

Hazard: Irritant

Formula: C₁₄H₁₈FNO₅
MDL Number: MFCD02682268
CAS Number: 221077-78-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR