N-Boc-3-cyano-L-phenylalanine

Supplier: Matrix Scientific

Synonyms: Boc-Phe(3-CN)-OH

041859-1G 041859-5G
101853-490EA 547.69 USD
101853-490 101853-492
N-Boc-3-cyano-L-phenylalanine
N-Boc-3-cyano-L-phenylalanine

Hazard: Irritant

Formula: C₁₅H₁₈N₂O₄
MW: 290.319 g/mol
MDL Number: MFCD00797560
CAS Number: 131980-30-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR