N-Boc-3-cyano-D-phenylalanine

Supplier: Matrix Scientific

Synonyms: Boc-D-Phe(3-CN)-OH

041858-1G 041858-5G
101853-486EA 547.69 USD
101853-486 101853-488
N-Boc-3-cyano-D-phenylalanine
N-Boc-3-cyano-D-phenylalanine

Hazard: Irritant

Formula: C₁₅H₁₈N₂O₄
MW: 290.319 g/mol
MDL Number: MFCD00797561
CAS Number: 205445-56-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR