Boc-3-chloro-L-tyrosine

Supplier: Matrix Scientific

Synonyms: Boc-Tyr(3-Cl)-OH

041857-1G 041857-5G
101853-482EA 661.54 USD
101853-482 101853-484
Boc-3-chloro-L-tyrosine
Boc-3-chloro-L-tyrosine

Hazard: Irritant

Formula: C₁₄H₁₈ClNO₅
MW: 315.76 g/mol
MDL Number: MFCD02682260
CAS Number: 192315-36-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR