Boc-3-chloro-L-phenylalanine

Supplier: Matrix Scientific

Synonyms: 3-Chloro-L-phenylalanine, N-BOC protected, Boc-Phe(3-Cl)-OH

041856-1G 041856-5G
101853-478EA 84.62 USD
101853-478 101853-480
Boc-3-chloro-L-phenylalanine
Boc-3-chloro-L-phenylalanine

Hazard: Irritant

Formula: C₁₄H₁₈ClNO₄
MW: 299.76 g/mol
MDL Number: MFCD00672515
CAS Number: 114873-03-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR