Boc-3-chloro-D-phenylalanine

Supplier: Matrix Scientific

041855-1G 041855-5G
101853-474EA 138.46 USD
101853-474 101853-476
Boc-3-chloro-D-phenylalanine
Formula: C₁₄H₁₈ClNO₄
MDL Number: MFCD00672516
CAS Number: 80102-25-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR